Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RanBP3 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1200000000000, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242672AF750

Catalog No. NP242672A70


Add to cart

Description

Description

RanBP3 Monoclonal antibody specifically detects RanBP3 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

RanBP3
1200000000000
Western Blot, ELISA
Conjugated, Purified
RANBP3
Recombinant human RANBP3 (235-445aa) purified from E. coli
Protein A purified
Signal Transduction
Primary
8498.0
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3
Mouse
IgG1 Kappa
100 μL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit