Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANBP3L Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548628
Description
Specifications
RANBP3L | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86VV4 | |
RANBP3L | |
KLH conjugated synthetic peptide between 137-165 amino acids from the Central region of human RANBP3L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RANBP3L | |
Ran-binding protein 3-like, RANBP3L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202151 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction