Learn More
RANBP9 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177551AP
Catalog No. 50-173-1768
Add to cart
Description
Description
This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.
Specifications
Specifications
RANBP9
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P69566, Q96S59
BPM L, BPM90, RAN binding protein 9, Ran binding protein M, RanBP7, RANBP9, RANBPM
Rabbit
IgG
150 μL
-20°C
Primary
10048, 364686, 478728, 56705
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RANBP9
Liquid
Ranbp9
RANBP9 Fusion Protein Ag12112
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Canine, Human, Mouse, Rat