Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANGAP1 Mouse anti-Canine, Human, Rat, Clone: OTI1B4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA501265
Description
RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1, 000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle.Specifications
RANGAP1 | |
Monoclonal | |
0.81 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P46060 | |
RANGAP1 | |
Full length human recombit protein of human RANGAP1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B4 | |
Unconjugated | |
RANGAP1 | |
Fug1, RANGAP, SD | |
Mouse | |
Affinity Chromatography | |
RUO | |
362965, 481234, 5905 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction