Learn More
Invitrogen™ RANGAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592435
Description
Immunogen sequence: PQQRGQGEKS ATPSRKILDP NTGEPAPVLS SPPPADVSTF LAFPSPEKLL RLGPKSSVLI AQQTDTSDPE KVVSAFLKVS SVFKDEATVR MAVQDAVDAL MQKAFNSSSF NSNTFLTRLL VHMGLLKSED KVKAIANLYG PLMALNHMVQ QDYFPKALAP LLLAFVTKPN SALESCSFAR HSLLQTLYKV; Positive Samples: BT-474, HeLa, MCF7, A-549, Jurkat; Cellular Location: Chromosome, Cytoplasm, Cytoplasmic side, Nucleus membrane, Peripheral membrane protein, centromere, cytoskeleton, kinetochore, spindle pole.
Ran is a small signaling GTPase that is involved in nucleocytoplasmic transport. Two additional functions of animal Ran in the formation of spindle asters and the reassembly of the nuclear envelope in mitotic cells have been recently reported. In contrast to Ras or Rho, Ran is not associated with membranes. Instead, the spatial sequestering of its accessory proteins, the Ran GTPase-activating protein RanGAP and the nucleotide exchange factor RCC1, appears to define the local concentration of RanGTP vs. RanGDP involved in signaling. Mammalian RanGAP is bound to the nuclear pore by a mechanism involving the attachment of small ubiquitin-related modifier protein (SUMO) to its C terminus and the subsequent binding of the SUMOylated domain to the nucleoporin Nup358.
Specifications
RANGAP1 | |
Polyclonal | |
Unconjugated | |
RANGAP1 | |
C79654; fug1; KIAA1835; MGC20266; mKIAA1835; RAN GTPase activating protein 1; RAN GTPase activating protein 1 a; Ran GTPase activating protein 1 b; Ran GTPase activating protein 1 L homeolog; Ran GTPase activating protein 1 S homeolog; ran GTPase-activating protein 1; RANGAP; RanGAP1; rangap1.L; rangap1.S; rangap1a; rangap1-a; rangap1b; rangap1-b; rna1p; SD; segregation distorter homolog; segregation distortion; XELAEV_18023679mg; XELAEV_18025680mg | |
Rabbit | |
Affinity chromatography | |
RUO | |
19387, 5905 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P46060, P46061 | |
RANGAP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 398-587 of human RanGAP1 (NP_0028741). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.