Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Ranger Insulated Safety Mid-Pac
Fleece-insulated, vulcanized rubber construction provides for warm, dry, comfortable feet
Supplier: Honeywell™ 6147RDM090
This item is not returnable.
View return policy
Description
Specialized compound prevents ozone deterioration for longer life footwear.
- Ultra-composite shank is longer, wider and positioned for optimal support to help prevent foot pain
- Insulated for added warmth in cold-weather applications
- Heavy Trac Tread sole and heel and knurled protective overlays are tough and durable
- Available in red, US men's sizes 7,9, 10-11, 13
- Height: 30.48cm (12 in.)
- Lace-up closure
- 100% waterproof
- Steel-toe
Compliance
Meets ASTM F2413-11 M I/75 C/75
Specifications
Red | |
12 in. | |
Rubber | |
9 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction