Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ranitidine hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
66357-59-3
C13H23ClN4O3S
350.862
GGWBHVILAJZWKJ-CHHCPSLASA-N
ranitidine hydrochloride,noctone,n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride,c13h22n4o3s.hcl,opera_id_624,melfax hydrochloride,zantac hydrochloride,zintac hydrochloride,azantac hydrochloride,raniben hydrochloride
6603542
(Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride
CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl
This item is not returnable. View return policy
66357-59-3
C13H23ClN4O3S
350.862
GGWBHVILAJZWKJ-CHHCPSLASA-N
ranitidine hydrochloride,noctone,n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride,c13h22n4o3s.hcl,opera_id_624,melfax hydrochloride,zantac hydrochloride,zintac hydrochloride,azantac hydrochloride,raniben hydrochloride
6603542
(Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride
CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl

Selective H2 antagonist

Potent, selective and competitive histamine H2 receptor antagonist (pA2 = 6.95 - 7.2). Inhibits gastric acid secretion induced by histamine, pentagastrin, bethanecol and food in vivo. Also inhibits aspirin-induced gastric lesions.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 350.86
Percent Purity >99%
Chemical Name or Material Ranitidine hydrochloride

Recommended Storage : Desiccate at +4°C