Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ranitidine Hydrochloride, USP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
66357-59-3
C13H23ClN4O3S
350.86
GGWBHVILAJZWKJ-KJEVSKRMSA-N
hydrogen dimethyl[(5-{[(2-{[(1E)-1-(methylamino)-2-nitroethenyl]amino}ethyl)sulfanyl]methyl}furan-2-yl)methyl]amine chloride
[H+].[Cl-].CN\C(NCCSCC1=CC=C(CN(C)C)O1)=C/[N+]([O-])=O
66357-59-3
C13H23ClN4O3S
350.86
GGWBHVILAJZWKJ-KJEVSKRMSA-N
hydrogen dimethyl[(5-{[(2-{[(1E)-1-(methylamino)-2-nitroethenyl]amino}ethyl)sulfanyl]methyl}furan-2-yl)methyl]amine chloride
[H+].[Cl-].CN\C(NCCSCC1=CC=C(CN(C)C)O1)=C/[N+]([O-])=O

R1719, 66357-59-3, C13H22N4O3S•HCl

Spectrum™ Ranitidine Hydrochloride, USP is used to prevent and treat heartburn and other symptoms caused by too much acid in the stomach. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.0075
pH 4.5 to 6.0
Ignition Residue 0.001
Quantity 5 g
CAS Max % 1
Percent Purity 97.5 to 102%
Grade USP