Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANK/TNFRSF11A Mouse anti-Human, Alexa Fluor 594, Clone: 80704, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB683T100UG
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human samples. It is validated for Flow Cytometry.Specifications
RANK/TNFRSF11A | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
Mouse myeloma cell line NS0-derived recombinant human RANK/TNFRSF11A extracellular domain Accession # Q9Y6Q6 | |
RUO | |
8792 | |
Human | |
IgG1 |
Flow Cytometry | |
80704 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y6Q6 | |
Mouse | |
100 μg | |
Primary | |
Detects human RANK/TNFRSF11A in ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhLymphotoxin beta R, rhNGF R, rhOPG, rhTAJ, rhTNF RI, rhTNF RII, or recombinant mouse (rm) OX40, and rmRANK is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)