Learn More
Selleck Chemical LLCSupplier Diversity Partner Ranolazine 100mg 95635-55-5 CVT 303, RS 43285-003  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1799100MG
Ranolazine (CVT 303, RS 43285-003) is a calcium uptake inhibitor via the sodium/calcium channel, used to treat chronic angina.
Catalog No. 50-661-3