Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1A, Rabbit anti-Human,Mouse,Rat,Porcine,Bovine,Canine,Chicken,Guinea Pig,Hamster,Primate,Rabbit,Sheep,Xenopus, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP197480AF488

Catalog No. NB004597

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RAP1A Polyclonal antibody specifically detects RAP1A in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot.
Specifications

Specifications

RAP1A
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
Rabbit
IgG
0.1 mL
Cell Cycle and Replication, Endosome Markers
Primary
Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus
Western Blot
Western Blot
Affinity Purified, Conjugated
RAP1A
Synthetic peptide corresponding to a portion of human Rap1a. The sequence is completely conserved in mouse, rat, bovine, chicken, Xenopus. Differs from a portion of Rap1b by 2 amino acids.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit