Learn More
Invitrogen™ RAP1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527580
Description
Recommended positive controls: HCT116, RAP1B-transfected 293T. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (97%), Xenopus laevis (99%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
Specifications
RAP1B | |
Polyclonal | |
Unconjugated | |
RAP1B | |
2810443E11Rik; cb1026; cb119; GTP-binding protein (smg p21B); GTP-binding protein smg p21B; K-REV; OK/SW-cl.11; RAL1B; rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; ras-related protein rab-1B-like protein; RAS-related protein RAP1B; ras-related protein Rap-1b; Ras-related protein Rap-1b-like protein; sb:cb119; si:dkey-31f5.4; small GTP binding protein; wu:fb11c04; wu:fb74e09; wu:fk70e05 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P61224 | |
RAP1B | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 156 of Human RAP1B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.