Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RAP1B |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6645
![]() ![]() |
Proteintech Group Inc
108401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
RAP1B and RAP1A belong to a superfamily of RAS like small GTP-binding proteins involved in cell signaling.Specifications
RAP1B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P61224, Q62636, Q99JI6 | |
171337, 215449, 5908 | |
RAP1B Fusion Protein Ag1282 | |
Primary | |
-20°C | |
RAP1B |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GTP binding protein smg p21B, K REV, RAL1B, RAP1B, Ras related protein Rap 1b | |
Rap1b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title