Learn More
RAP1GDS1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00005910T01
Catalog No. 89-001-969
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_021159.3
6.8
Transient overexpression cell Lysate was tested with Anti-RAP1GDS1 antibody (H00005910-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
RAP1GDS1 293T Cell Transient Overexpression Lysate(Denatured)
66.3
pCMV-RAP1GDS1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.