Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAP80 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520413
Description
A suggested positive control is 293 cell lysate. PA5-20413 can be used with blocking peptide PEP-0530.
RAP80 was initially identified as zinc-finger containing nuclear protein that is highly expressed in testis and interacts with the retinoid-related testis-associated receptor (RTR). Later experiments revealed that RAP80 is recruited by the Coiled-coil domain 98 (CCDC98) protein to the breast cancer-1 protein BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both RAP80 and CCDC98 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either RAP80 or CCDC98 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that RAP80 and CCDC98 control only part of the DNA damage response role of BRCA1. At least four isoforms of RAP80 are known to exist.
Specifications
RAP80 | |
Polyclonal | |
Unconjugated | |
UIMC1 | |
9430016E08Rik; BRCA1-A complex subunit RAP80; D330018D10Rik; D630032M02Rik; RAP80; Receptor-associated protein 80; retinoid X receptor interacting protein 110; retinoid X receptor-interacting protein 110; RGD1307009; RIP110; Rxrip110; ubiquitin interaction motif containing 1; ubiquitin interaction motif-containing protein 1; Uimc1; X2HRIP110 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20184, 51720 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5U5Q9, Q96RL1 | |
UIMC1 | |
A 17 amino acid peptide from near the carboxy terminus of human RAP80. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction