Learn More
RAPGEF1 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI2C3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA526281
Catalog No. PIMA526281
Add to cart
Description
Description
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
Specifications
Specifications
RAPGEF1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13905
GRF2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
107746, 2889, 491286, 63881, 715272
Immunohistochemistry (Paraffin), Western Blot
OTI2C3
Unconjugated
RAPGEF1
Liquid
Rapgef1
Full length human recombit protein of RAPGEF1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat