Learn More
Abnova™ RAPGEF5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00009771T02
Catalog No. 89-959-376
Add to cart
Description
Description
Specifications
Specifications
293T
BC039203
pCMV-RAPGEF5 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
RAPGEF5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.95kDa
Transient overexpression cell lysate was tested with anti-RAPGEF5 antibody (Mfr. No. H00009771-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human