Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550106

Catalog No. PA550106


Add to cart

Description

Description

The antibody detects endogenous levels of total RAPGEF5 protein.

Guanine nucleotide exchange factor (GEF) for RAP1A, RAP2A and MRAS/M-Ras-GTP. Its association with MRAS inhibits Rap1 activation. Nagase T., DNA Res. 3:321-329(1996). Scherer S.W., Science 300:767-772(2003). Ichiba T., FEBS Lett. 457:85-89(1999)
Specifications

Specifications

RAPGEF5
Polyclonal
Unconjugated
RAPGEF5
Liquid
RAPGEF5
Synthesized peptide derived from internal of human RAPGEF5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q92565, Q8C0Q9
GFR, KIAA0277, MRGEF
Rabbit
IgG
100 μL
-20°C
Primary
217944, 9771
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit