Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259126

Catalog No. NBP259126


Add to cart

Description

Description

RAPH1 Polyclonal antibody specifically detects RAPH1 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

RAPH1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
candidate 18, candidate 9, lamellipodin, PREL2, PREL-2, proline rich EVH1 ligand 2, Protein RMO1, Ras association (RalGDS/AF-6) and pleckstrin homology domains 1, ras-associated and pleckstrin homology domains-containing protein 1, RMO1
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Immunoprecipitation
Immunoprecipitation
Affinity Purified
RAPH1
Maps to a region between 1200 and 1250
Immunogen affinity purified
RUO
Primary
65059
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only