Learn More
Cell Biolabs IncSupplier Diversity Partner RAPID ANTIBODY PURIFICATION KT  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner AKR160
The Rapid Antibody Purification Kit produces high quality IgG by Protein A column. Suitable for use with ascites, serum and tissue culture media or hybridoma supernatants. 10 preps/kit.
Catalog No. 50-672-579