Learn More
Cliawaived Inc Rapid TSH Cassettes
SDP
Manufacturer:  Cliawaived Inc CLIATSH120A
Encompass_Preferred
(20/Pack) CLIAwaived, Inc. Rapid TSH test for Hypothyroidism. The TSH test is qualitative test for elevated thyroid stimulating hormone (TSH) in whole blood. Requires only one drop of blood from a finger stick or venous draw. Results in 10 min. FDA-cleared and CLIAwaived. (FDA 510K#: K990658)Controls NOT included.
Catalog No. NC1426364