Learn More
Thermo Scientific RapidStak Instrument Drivers
Easily integrate a wide variety of lab instruments and devices into a Polara RS automated lab system using Thermo Scientific RapidStak Instrument Drivers.
Manufacturer: Thermo Scientific RSI0001
Description
Easily integrate a wide variety of lab instruments and devices into a Polara RS automated lab system using Thermo Scientific RapidStak Instrument Drivers. Polara RS Instrument drivers support instrument pooling, enabling you to add instruments for higher throughput without having to change your methods. You never have to specify robot operations in your method; Polara RS and Polara RS Instrument drivers determine those for you.
Works with the Following Thermo Scientific Instruments:
- Multidrop Combi
- Multidro Combi nL
- Multidrop 384
- Wellwash Versa
- Multiskan GO
- Multiskan FC
- Varioskan Flash
- SkanIt readers
- Ascent readers
- ALPS 3000
- ALPS 300
- Seal-It 100
Specifications
Polara RS new instrument interface | |
Rapid Stak Instrument Driver |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.