Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RARalpha/NR1B1 Rabbit anti-Human, Clone: SN0725, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP267712

Catalog No. NBP267712


Add to cart

Description

Description

RARalpha/NR1B1 Monoclonal antibody specifically detects RARalpha/NR1B1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

RARalpha/NR1B1
SN0725
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha
Rabbit
IgG
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Flow Cytometry, Western Blot
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:50-1:100
Purified
RARA
N terminal human Retinoic Acid Receptor alpha
Protein A purified
Cancer, Phospho Specific, Vision
Primary
5914
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit