Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RARRES1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550585
Description
The antibody detects endogenous levels of total TIG1 protein.
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed.
Specifications
RARRES1 | |
Polyclonal | |
Unconjugated | |
RARRES1 | |
5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5918 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P49788 | |
RARRES1 | |
Synthetic peptide corresponding to a region derived from internal residues of human Retinoic acid receptor responder (tazarotene induced) 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction