Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RARRES1 Monoclonal Antibody (OTI1B3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526246
Description
RARRES1 Monoclonal antibody specifically detects RARRES1 in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RARRES1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49788 | |
RARRES1 | |
Full length human recombinant protein of RARRES1 produced in HEK293T cell | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
OTI1B3 | |
Unconjugated | |
RARRES1 | |
5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
109222, 310486, 5918, 612298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction