Learn More
RARRES3 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6139SP
Catalog No. AF6139SP
Add to cart
This item is not returnable. View return policy
Description
Description
RARRES3 Polyclonal antibody AF6139SP specifically detects RARRES3 in Human samples. It is validated for Immunocytochemistry
Specifications
Specifications
RARRES3
LYOPH
Immunocytochemistry 5-15 ug/mL
Q9UL19
HRASLS4, PLA1/2-3, RAR-responsive protein TIG3, retinoic acid receptor responder (tazarotene induced) 3, retinoic acid receptor responder protein 3, retinoic acid-inducible gene 1, Retinoid-inducible gene 1 protein, RIG1, Tazarotene-induced gene 3 protein, TIG3, TIG3MGC8906
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human RARRES3 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) RARRES1 and rhLatexin is observed.
Human
Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RARRES3
E. coli-derived recombinant human RARRES3 Asp12-Lys134 Accession # Q9UL19
Immunogen affinity purified
RUO
Primary
5920
Reconstitute at 0.2 mg/mL in sterile PBS.