Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RARS Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519236
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.
Specifications
RARS | |
Polyclonal | |
Unconjugated | |
RARS1 | |
2610011N19Rik; 2610037E21Rik; AL033339; arginine tRNA ligase 1, cytoplasmic; arginine--tRNA ligase, cytoplasmic; Arginyl-tRNA synthetase; arginyl-tRNA synthetase, cytoplasmic; argRS; AW985894; DALRD1; HLD9; MGC8641; RARS | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5917 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P54136 | |
RARS1 | |
Synthetic peptide sequence (SLQAERNKPTKN) corresponding to the internal amino acids of RARS (aa 61-73). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction