Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Ras, Mouse, Alexa Fluor 647, Clone: RAS10,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 05516AF647
Description
Ras proteins are small GTPases controlling cellular pathways that signal for growth, proliferation, and differentiation. Mammalian genomes harbor three Ras genes (HRAS, NRAS, and KRAS) that encode four GTPases, two of which (K-Ras4A and K-Ras4B) are transcribed from the KRAS gene via two alternative fourth exons. Ras proteins share nearly identical sequence homology in sequence encoded by exons 1-3 (a.a. 1-165), while diverge in their C-terminal 24-a.a. hypervariable region (HVR) encoded by exon 4. The first 165 amino acids form the catalytic G domain that mediates GDP/GTP binding and association with effectors, exchange factors, and GTPase-activating proteins (GAPs), while the C-terminal HVR is needed for Ras membrane localization, a process necessary for Ras function. In addition to the removal of initiator methionine (Met1), Ras proteins are posttranslationally modified by farnesylation of Cys186, followed by proteolytical cleavage their C-terminal end three amino acids (a.a. 187-189) and carboxyl methylation of the newly formed C-terminal prenylcysteine. H-Ras, N-Ras, and K-Ras4A are further palmitoylated on cysteine residue(s) near the C-terminus (Cys181/Cys184 in H-Ras, Cys181 in N- and K-Ras), while K-Ras4B lacks a palmitoylation site. Palmitoylation is required for targeting H- and N-Ras to the plasma membrane (PM), while K-Ras4A and K-Ras4B contain a series of lysines that constitute a polybasic region (PBR) capable of interacting with the inner leaflet of the PM through an electrostatic interaction.Specifications
Ras | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse IgG2a Alexa Fluor™ 647 conjugate in buffer containing PBS with 1.5% BSA and 0.05% Sodium Azide. | |
P01112 | |
Recombinant human H-Ras p21 (Hamer, P.J., et al. (1990). Hybridoma.9(6):573-587). | |
100 μL | |
Signaling | |
NM_001130442;NM_005343 | |
Human | |
Purified |
Immunocytochemistry | |
RAS10 | |
Alexa Fluor 647 | |
HRAS, HRAS1, KRAS, CFC2, NS3, KRAS2, RASK2, NRAS, NS6, CMNS, NCMS | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Ras 10 reacts with normal p21, activated p21, and p21 from each of the Harvey, Kirsten and N-ras families. Ras 10 reacts with recombinant p21 as well as p21 derived from human and rodent cells (Hamer, P.J., et al. (1990). Hybridoma.9(6):573-587). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction