Learn More
Ras Rat anti-Human, Alexa Fluor™ 532, Clone: MM0522-8F13, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212433AF532
Catalog No. NP212433A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ras Monoclonal antibody specifically detects Ras in Human samples. It is validated for Western Blot.
Specifications
Specifications
Ras
MM0522-8F13
Western Blot
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Human
0.1 mL
Primary
Human