Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RASD2 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $976.00
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl (pH8.0) containing 10% glycerol |
Gene ID (Entrez) | 23551 |
Molecular Weight | TMW: 32.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25198101
|
Novus Biologicals™
NBP2519810.1MG |
0.1mg |
Each for $344.00
|
|
NBP25198105
|
Novus Biologicals™
NBP2519810.5MG |
0.5mg |
Each for $976.00
|
|
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-266 of Human RASD2 The Recombinant Human RASD2 Protein is derived from E. coli. The Recombinant Human RASD2 Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
20mM Tris-HCl (pH8.0) containing 10% glycerol | |
TMW: 32.8kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
RASD2 | |
Yes |
SDS-Page | |
23551 | |
>85%, by SDS-PAGE | |
GTP-binding protein Rhes, MGC:4834, RASD family, member 2, Rhes, TEM2Ras homolog enriched in striatum, Tumor endothelial marker 2 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title