Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGEF1C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548914

Catalog No. PA548914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Guanine nucleotide exchange factor (GEF).
Specifications

Specifications

RASGEF1C
Polyclonal
PBS with 0.09% sodium azide
Q8N431
Ras-GEF domain-containing family member 1C, RASGEF1C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
255426
Western Blot
Unconjugated
RASGEF1C
Liquid
RASGEF1C
KLH conjugated synthetic peptide between 79-105 amino acids from the N-terminal region of human RASGEF1C.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit