Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASGRF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115827
Description
Antibody detects endogenous levels of total RASGRF2.
RAS (MIM 190020) GTPases cycle between an inactive GDP-bound state and an active GTP-bound state. Guanine-nucleotide exchange factors (GEFs), such as RASGRFs, stimulate the conversion of the GDP-bound form into the active form.
Specifications
RASGRF2 | |
Polyclonal | |
Unconjugated | |
RASGRF2 | |
6330417G04Rik; AW048350; Grf2; guanine nucleotide-releasing factor 2; ras guanine nucleotide exchange factor 2; Ras protein specific guanine nucleotide releasing factor 2; RAS protein-specific guanine nucleotide-releasing factor 2; Rasgrf2; ras-GRF2; Ras-specific guanine nucleotide-releasing factor 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
114513, 19418, 5924 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O14827, P70392, Q99JE4 | |
RASGRF2 | |
A synthesized peptide derived from human RASGRF2(Accession O14827), corresponding to amino acid residues F54-G104. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction