Learn More
RASGRP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 197451AP
Catalog No. 50-173-2522
Add to cart
Description
Description
The antibody recognizes the C-term of RASGRP2.
The protein encoded by this gene is a brain-enriched nucleotide exchanged factor that contains an N-terminal GEF domain, 2 tandem repeats of EF-hand calcium-binding motifs, and a C-terminal diacylglycerol/phorbol ester-binding domain. This protein can activate small GTPases, including RAS and RAP1/RAS3. The nucleotide exchange activity of this protein can be stimulated by calcium and diacylglycerol. Three alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
RASGRP2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7LDG7, Q9QUG9
CALDAG GEFI, Cdc25 like protein, CDC25L, F25B3.3 kinase like protein, hCDC25L, MCG7, RAS guanyl releasing protein 2, RASGRP2
Rabbit
IgG
150 μL
-20°C
Primary
10235, 19395
Western Blot
Unconjugated
RASGRP2
Liquid
RASGRP2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse