Learn More
RASGRP3 Mouse anti-Human, Clone: OTI3C5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801445
Catalog No. 50-166-5599
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
Specifications
Specifications
RASGRP3
Monoclonal
Unconjugated
RASGRP3
Lyophilized
RASGRP3
Human recombit protein fragment corresponding to amino acids 380-690 of human RASGRP3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3C5
PBS with 8% trehalose and no preservative; pH 7.3
Q8IV61
GRP3
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25780