Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRP4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521597

Catalog No. PIPA521597


Only null left
Add to Cart

Description

Description

Recommended positive controls: Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RASGRP4
Polyclonal
Unconjugated
RASGRP4
guanyl nucleotide releasing protein 4; RAS guanyl releasing protein 4; RAS guanyl-releasing protein 4; RASGRP4
Rabbit
Antigen affinity chromatography
RUO
115727
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q8TDF6
RASGRP4
Synthetic peptide corresponding to a region within amino acids 108 and 200 of RASGRP4 (Uniprot ID#Q8TDF6).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.