Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASL10A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563255
Description
Immunogen sequence: AKYNWHVLRL FRELLRCALV RARPAHPALR LQGALHPARC SLM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 98%.
The specific function of RASL10A is not yet known.
Specifications
RASL10A | |
Polyclonal | |
Unconjugated | |
RASL10A | |
2210403B10Rik; AI852688; RAS like family 10 member A; RASL10A; RAS-like family 10 member A; ras-like protein family member 10A; Ras-like protein RRP22; Ras-like protein RRP22 (RAS-related protein on chromosome 22); RAS-like, family 10, member A; ras-related protein on chromosome 22; Ras-related protein on chromosome 22 homolog; RGD1306100; RRP22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10633 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92737 | |
RASL10A | |
Recombinant protein corresponding to Human RASL10A. Recombinant protein control fragment (Product #RP-101152). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction