Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASL11B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549640

Catalog No. PA549640

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse, rat and xenopus based on sequence homology.

RASL11B is a member of the small GTPase protein family with a high degree of similarity to RAS (see HRAS, MIM 190020) proteins.[supplied by OMIM, Nov 2008]
Specifications

Specifications

RASL11B
Polyclonal
PBS with 0.09% sodium azide
Q9BPW5
Ras-like protein family member 11B, RASL11B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65997
Flow Cytometry, Western Blot
Unconjugated
RASL11B
Liquid
RASL11B
KLH conjugated synthetic peptide between 37-66 amino acids from the N-terminal region of human RASL11B.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit