Learn More
Invitrogen™ RASL12 Monoclonal Antibody (1C10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524464
Description
Peptide Sequence: MSSVFGKPRA GSGPQSAPLE VNLAILGRRG AGKSALTVEF LTKRFISEYD PNLEDTYSSE ETVDHQPVHL RVMDTADLDT PRNCERYLNW AHAFLVVYSV DSRQSFDSSS SYLELLALHA KETQRSIPAL LLGNKLDMAQ YRQVTKAEGV ALAGRFGCLF FEVSACLDFE HVQHVFHEAV REARRELEKS PLTRPLFISE ERALPHQAPL TARHGLASCT FDTLSTINLK EMPTVAQAKL VTVKSSRAQS KRKAPTLTLL KGFKIF.
RASL12 belongs to the small GTPase superfamily. Ras family.
Specifications
RASL12 | |
Monoclonal | |
Unconjugated | |
RASL12 | |
4631404I11Rik; LOC100714151; Ras family member Ris; RAS like family 12; Rasl12; RAS-like family 12; ras-like protein family member 12; Ras-like protein Ris; RAS-like, family 12; RAS-like, family 12 protein; RIS | |
Mouse | |
Protein A | |
RUO | |
51285 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
1C10 | |
PBS with no preservative; pH 7.4 | |
Q9NYN1 | |
RASL12 | |
RASL12 (AAH53734, 1 a.a. ~ 266 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.