Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF10 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548377

Catalog No. PA548377

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RASSF10
Polyclonal
PBS with 0.09% sodium azide
A6NK89, Q8BL43
Ras association domain-containing protein 10, RASSF10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
644943, 78748
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RASSF10
Liquid
RASSF10
KLH conjugated synthetic peptide between 230-259 amino acids from the Central region of human RASSF10.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit