Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550869

Catalog No. PA550869


Add to cart

Description

Description

The antibody detects endogenous levels of total RASSF3 protein.

The RAS oncogene is mutated in nearly one-third of all human cancers. Members of the RAS superfamily are plasma membrane GTP-binding proteins that modulate intracellular signal transduction pathways. A subfamily of RAS effectors, including RASSF3, share a RAS association domain.
Specifications

Specifications

RASSF3
Polyclonal
Unconjugated
RASSF3
Liquid
RASSF3
Full length fusion protein of Human RASSF3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86WH2
RASSF5
Rabbit
IgG
100 μL
-20°C
Primary
283349
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit