Learn More
Invitrogen™ RASSF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113594
Description
Immunogen sequence: QSAEVQGSLA LVSRALEAAE RALQAQAQEL EELNRELRQC NLQQFIQQTG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%; Yeast: 79%.
Negatively regulates stress-induced JNK activation and apoptosis by promoting MAP2K7 phosphorylation and inhibiting its ability to activate JNK. Following prolonged stress, anti-apoptotic effect stops because of degradation of RASSF7 protein via the ubiquitin-proteasome pathway. Required for the activation of AURKB and chromosomal congression during mitosis where it stimulates microtubule polymerization. [UniProt].
Specifications
RASSF7 | |
Polyclonal | |
Unconjugated | |
RASSF7 | |
2400009B11Rik; AW210608; C11orf13; HRAS1; HRAS1-related cluster protein 1; HRAS1-related cluster-1; HRC1; LOW QUALITY PROTEIN: ras association domain-containing protein 7; MGC126069; MGC126070; Ras association (RalGDS/AF-6) domain family (N-terminal) member 7; Ras association (RalGDS/AF-6) domain family 7; Ras association domain family member 7; ras association domain-containing protein 7; Rassf7; RGD1306244 | |
Rabbit | |
Affinity chromatography | |
RUO | |
8045 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q02833 | |
RASSF7 | |
Synthetic peptide directed towards the middle region of human RASSF7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.