Learn More
Novus Biologicals™ Rat 11-dehydro-TXB2 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266721
Catalog No. NBP266721
Add to Cart
Description
Description
The Rat 11-dehydro-TXB2 ELISA Kit (Colorimetric) measures rat 11-dehydro-TXB2 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Rat 11-DH-TXB2 in samples. No significant cross-reactivity or interference between Rat 11-DH-TXB2 and analogues was observed.
Biological Fluids, Plasma, Serum
Rat
Storage of components varies. See protocol for specific instructions.
9.38 pg/mL
ELISA Kit (Colorimetric)
ELISA
1 Kit
11-dehydro-TXB2
Recommendations
Recommendations