Learn More
R&D Systems™ Rat 4-1BB Ligand/TNFSF9 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$280.50
Specifications
Product Type Recombinant protein
Regulatory Status RUO
Recombinant Yes
Biological Activity 5 to 30ng/mL
Quantity 25μg
View More Specs
Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
91-884-L025 View Documents R&D Systems™
91884L025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein Each for $280.50
Add to cart
91-884-L025CF View Documents R&D Systems™
91884L025/CF
Lyophilized from a 0.2μm filtered solution in PBS Each for $280.50
Add to cart
Description
Description
The Recombinant Rat 4-1BB Ligand/TNFSF9 Protein is derived from NS0. The Recombinant Rat 4-1BB Ligand/TNFSF9 Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Recombinant protein
Yes
25μg
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<1.0 EU per 1μg of the protein by the LAL method
Mouse myeloma cell line, NS0-derived Arg103-Gly308, with an N-terminal 6-His tag
RUO
5 to 30ng/mL
Unlabeled
Rat
24 kDa
Q80WE6