Learn More
Novus Biologicals™ Rat AHR ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP266671
Description
The Rat AHR ELISA Kit (Chemiluminescence) measures rat AHR in serum, plasma and other biological fluids.Specifications
62.50-4000 pg/mL | |
This kit recognizes Rat AhR in samples. No significant cross-reactivity or interference between Rat AhR and analogues was observed. | |
196 | |
Rat | |
1 Kit | |
Storage is content dependent. |
37.50 pg/mL | |
ELISA | |
AHR | |
ELISA Kit (Chemiluminescence) | |
Biological Fluids,Plasma,Serum | |
AHR |