Learn More
Novus Biologicals™ Rat AIF-1/Iba1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266675
Catalog No. NBP266675
Add to cart
Description
Description
The Rat AIF-1/Iba1 ELISA Kit (Colorimetric) measures rat AIF-1/Iba1 in serum, plasma and other biological fluids.
Specifications
Specifications
Biological Fluids, Plasma, Serum
31.25 - 2000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat 1AIF1 in samples. No significant cross-reactivity or interference between Rat 1AIF1 and analogues was observed.
199
18.75 pg/mL
AIF-1/Iba1
AIF1