Learn More
Novus Biologicals™ Rat AIP/ARA9 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP266680
Description
The Rat AIP/ARA9 ELISA Kit (Colorimetric) measures rat AIP/ARA9 in serum, plasma and other biological fluids.Specifications
1.56 - 100 ng/mL | |
This kit recognizes Rat AIP in samples. No significant cross-reactivity or interference between Rat AIP and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
9049 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.94 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
AIP | |
1 Kit | |
AIP/ARA9 |