Learn More
Invitrogen™ Rat Angiopoietin-1 ELISA Kit
ELISA
$700.00 - $5890.00
Specifications
Accession Number | 1110046O21Rik; AGP1; AGPT; Agpt1; Ang1; Ang-1; angiopoietin 1; angiopoietin-1; ANGPT1; endothelial growth factor; KIAA0003 |
---|---|
Assay Range | 0.4 to 100 ng/mL |
Assay Sensitivity | 0.4 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ERA3RBX5
![]() |
Invitrogen™
ERA3RBX5 |
5 x 96 Tests |
Each for $3,285.00
|
|
|||||
ERA3RB
![]() |
Invitrogen™
ERA3RB |
96 Tests |
Each for $700.00
|
|
|||||
50-254-5432
![]() |
Invitrogen™
ERA3RBX10 |
10 x 96 Tests |
Each for $5,890.00
|
|
|||||
Description
Rat Angiopoietin-1 quantitates rat Angiopoietin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Angiopoietin-1.
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.Specifications
1110046O21Rik; AGP1; AGPT; Agpt1; Ang1; Ang-1; angiopoietin 1; angiopoietin-1; ANGPT1; endothelial growth factor; KIAA0003 | |
0.4 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Angpt1 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
Angiopoietin 1 | |
4 hrs 45 mins |
0.4 to 100 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
89807 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.