Learn More
Aflatoxin, Rat, Clone: IC6, Abnova™
Rat monoclonal antibody raised against Aflatoxin.
Manufacturer:  Abnova Corporation MAB6371
Catalog No. 89-136-194
Add to cart
Description
Description
Specifications
Specifications
Aflatoxin
IC6
Rat monoclonal antibody raised against Aflatoxin.
Lyophilized from 10mM PBS, pH 7.2
Rat
IgG2b, kappa
RUO
Primary
This antibody is reactive to Aflatoxin M1 (AFM1) and Aflatoxin B1 (AFB1), but not reactive to BSA and other irrelevant antigens by ELISA.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Lyophilized
Winstar rats were immunized with Aflatoxin M1 conjugated with BSA.
500 ug
Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Monoclonal