Learn More
Rat anti-Aminopeptidase A/ENPEP, Clone: 6C3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1432810.5MG
Catalog No. NBP1432815
Add to cart
Description
Description
Aminopeptidase A/ENPEP Monoclonal antibody specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
Aminopeptidase A/ENPEP
6C3
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Affinity Purified
ENPEP
The immunogen for this antibody was Aminopeptidase A/APA.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Q07075
Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160
Rat
IgG2a Kappa
0.5 mg
Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
2028