Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2 Rat anti-Mouse, Brilliant Violet 421, Clone: BZ2E3, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 565207

Catalog No. BDB565207


Add to cart

Description

Description

The BZ2E3 monoclonal antibody specifically binds to CCRL2 (C-C chemokine receptor-like 2) which is also known as Lipopolysaccharide-inducible C-C chemokine receptor (L-CCR/Lccr). CCRL2 is a chemokine receptor-like, 7-transmembrane protein that can bind CCL19 or the nonchemokine chemoattractant, chemerin/RARRES2. CCRL2 is expressed by a variety of cell types including mast cells, macrophages and some endothelial cells. CCRL2 appears to play various roles in regulating chemokine activities, e.g., by binding and internalizing excess local levels of the CCL19 /MIP-3β chemokine. Alternatively, endothelial cell-expressed CCRL2 can bind and present the chemerin/RARRES2 chemoattractant to lymphoid cells that express the functional signaling receptor, CMKLR1, and promote their adhesion.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CCRL2
BZ2E3
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
C-C chemokine receptor-like 2; L-CCR; Lccr; Cmkbr1l2; Ackr5
Affinity Purified
Mouse CCRL2 Peptide
Affinity Chromatography
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit